1,2-Dibromo-3-chloropropane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Chloro-2,3-dibromopropane
M-Clarity™ quality level = MQ100
676713-25G 676713-5G
SIAL676713-25GEA 99.9 EUR
SIAL676713-25G SIAL676713-5G
1,2-Dibromo-3-chloropropane, Sigma-Aldrich®
1,2-Dibromo-3-chloropropane
Formula: ClH₂CBrCHCH₂Br
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: 6.1 °C
Density: 2.085 g/cm³ (9 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039365
CAS Number: 96-12-8
EINECS: 202-479-3
UN: 2872
ADR: 6.1,III
Merck Index: 12,03068