5-Bromovaleryl chloride, Sigma-Aldrich®

Supplier: Merck

331317-25G 331317-5G
SIAL331317-25GEA 93.5 EUR
SIAL331317-25G SIAL331317-5G
5-Bromovaleryl chloride, Sigma-Aldrich®
5-Bromovaleryl chloride
Formula: BrH₂C(CH₂)₃COOCl
MDL Number: MFCD00013660
CAS Number: 4509-90-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR