4-Bromo-1-butanol, Technical Grade
Supplier: Apollo Scientific
Synonyms: 1-Bromo-4-hydroxybutane
OR4233-5G OR4233-25G
APOSOR4233-5GEA 69.6 EUR
APOSOR4233-5G APOSOR4233-25G
4-Bromo-1-butanol, Technical Grade
4-Bromo-1-butanol
Formula: BrH₂C(CH₂)₂CH₂OH
MW: 153.02 g/mol
Boiling Pt: 63…65 °C (5 mm Hg)
Density: 1.68 g/cm³
Flash Pt: 7 °C
Storage Temperature: Freezer
MDL Number: MFCD00015387
CAS Number: 33036-62-3
EINECS: 251-355-5
UN: 1987
ADR: 3 III