1,2-Epoxyhexane 96%
Supplier: Thermo Fisher Scientific
|
Danger
|
L07609.14
L07609.22
L07609.14EA
50
EUR
L07609.14
L07609.22
1,2-Epoxyhexane 96%
1,2-Epoxyhexane
Purity: 96%
CAS No.: 1436-34-6
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 15°(59°F)
Refractive Index: 1.4060
Boiling Point: 118-120°
Beilstein Reference: 102568
UN No.: 1993
MDL No.: MFCD00005154
Formula:
C₆H₁₂O Boiling Pt: 118…120 °C Density: 0.832 Flash Pt: 15 °C (59 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005154 CAS Number: 1436-34-6 EINECS: 215-864-6 UN: 1993 ADR: 3,II |