Propylene oxide
Supplier: Roth Carl
Danger
8617.1 8617.2 8617.3
ROTH8617.1EA 0 EUR
ROTH8617.1 ROTH8617.2 ROTH8617.3
Propylene oxide
Propylene oxide
Formula: C₃H₆O
Boiling Pt: 34.3 °C (1013 hPa)
Melting Pt: –112 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: –37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005126
CAS Number: 75-56-9
EINECS: 200-879-2
UN: 1280
ADR: 3,I
Merck Index: 12,08041
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=443494ff0620e18a4c5de7f3f4cd87d6e5eec44c&vid=33bc95a664261fbc904256d1b87a1972dde5dcd1&prid=web&clid=SAPDAM