Heptanal, Sigma-Aldrich®

Supplier: Merck
Danger

H2120-100ML H2120-1L H2120-2ML H2120-500ML
SIALH2120-100MLEA 44.4 EUR
SIALH2120-100ML SIALH2120-1L SIALH2120-2ML SIALH2120-500ML
Heptanal, Sigma-Aldrich®
Heptanal
Formula: C₇H₁₄O
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –43 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00007028
CAS Number: 111-71-7
EINECS: 203-898-4
UN: 3056
ADR: 3,III
Merck Index: 13,04677

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR