1-Chlorobutane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: n-Butyl Chloride
M-Clarity™ quality level = MQ100
414255-100ML 414255-1L
SIAL414255-100MLEA 74.8 EUR
SIAL414255-100ML SIAL414255-1L
1-Chlorobutane, Sigma-Aldrich®
1-Chlorobutane
Formula: CH₃(CH₂)₃Cl
MW: 92.57 g/mol
Boiling Pt: 78.4
Melting Pt: –123 °C
Density: 0.8862 g/cm³ (20 °C)
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00001009
CAS Number: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3 II
Merck Index: 13,01559
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bd980eb6e7ecd91951d46d0ea510119e976dc0&vid=5231549172a609ad96623a80204b11c7c0c69273&prid=web&clid=SAPDAM