1-Chlorodecane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
C32909-100G
C32909-500G
SIALC32909-100GEA
136
EUR
SIALC32909-100G
SIALC32909-500G
1-Chlorodecane, Sigma-Aldrich®
1-Chlorodecane
Formula:
C₁₀H₂₁Cl Boiling Pt: 223 °C (1013 hPa) Melting Pt: –34 °C Density: 0.868 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000956 CAS Number: 1002-69-3 EINECS: 213-691-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...