1-Chlorohexane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
238465-100ML 238465-500ML
SIAL238465-100MLEA 47 EUR
SIAL238465-100ML SIAL238465-500ML
1-Chlorohexane, Sigma-Aldrich®
1-Chlorohexane
Formula: C₆H₁₃Cl
Boiling Pt: ∼135 °C (1013 hPa)
Melting Pt: –94 °C
Density: 0.872 g/cm³ (20 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00001018
CAS Number: 544-10-5
EINECS: 208-859-5
UN: 1993
ADR: 3,III
Merck Index: 13,02163