2-Chlorobutane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
C28898-100ML C28898-250ML
SIALC28898-100MLEA 29.2 EUR
SIALC28898-100ML SIALC28898-250ML
2-Chlorobutane, Sigma-Aldrich®
2-Chlorobutane
Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 68
Melting Pt: –132 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: –10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000871
CAS Number: 78-86-4
EINECS: 201-151-7
UN: 1127
ADR: 3 II
Merck Index: 13,01560