5-Chloro-2-pentanone, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
C62603-100G C62603-25G
SIALC62603-100GEA 159 EUR
SIALC62603-100G SIALC62603-25G
5-Chloro-2-pentanone, Sigma-Aldrich®
5-Chloro-2-pentanone
Formula: C₅H₉ClO
Boiling Pt: 71 °C (1013 hPa)
Melting Pt: –48 °C
Density: 1.057 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001008
CAS Number: 5891-21-4
EINECS: 227-565-8
UN: 1224
ADR: 3,III