Chloral hydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2,2,2-Trichloroethane-1,1-diol
15307-100G-R 15307-2.5KG-R 15307-500G-R 15307-6X2.5KG- 15307-6X500G-R
SIAL15307-100G-REA 46.8 EUR
SIAL15307-100G-R SIAL15307-2.5KG-R SIAL15307-500G-R SIAL15307-6X2.5KG- SIAL15307-6X500G-R
Chloral hydrate, Sigma-Aldrich®
Chloral hydrate
Formula: Cl₃CCH(OH)₂
MW: 165.4 g/mol
Boiling Pt: 97.5
Melting Pt: 52
Density: 1.901 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044479
CAS Number: 302-17-0
EINECS: 206-117-5
UN: 2811
ADR: 6.1 III
Merck Index: 12,02113
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5db96e6ffe14849c6ffd0c2edd3db048fe472e17&vid=ca79a0593c65b3105b1880795fdc5b4255a7a421&prid=web&clid=SAPDAM