2-Nonanone, Sigma-Aldrich®
Supplier: Merck
Synonyms: Methyl heptyl ketone, Heptyl methyl ketone
M-Clarity™ quality level = MQ100
108731-100G 108731-5G
SIAL108731-100GEA 64.4 EUR
SIAL108731-100G SIAL108731-5G
2-Nonanone, Sigma-Aldrich®
2-Nonanone
Formula: C₉H₁₈O
MW: 142.24 g/mol
Boiling Pt: 192
Melting Pt: –15 °C
Density: 0.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009553
CAS Number: 821-55-6
EINECS: 212-480-0