2-Pentanone, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Methyl Propyl Ketone
M-Clarity™ quality level = MQ300
W284203-100G W284203-1KG W284203-20KG W284203-4KG W284203-SAMPLE
SIALW284203-100GEA 68.4 EUR
SIALW284203-100G SIALW284203-1KG SIALW284203-20KG SIALW284203-4KG SIALW284203-SAMPLE
2-Pentanone, Sigma-Aldrich®
2-Pentanone
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 100
Melting Pt: –78 °C
Density: 0.809 g/cm³ (20 °C)
Flash Pt: 7
Storage Temperature: Ambient
MDL Number: MFCD00009400
CAS Number: 107-87-9
EINECS: 203-528-1
UN: 1249
ADR: 3 II
Merck Index: 12,06193