Methyl ethyl ketone, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK
M-Clarity™ quality level = MQ300
W217018-1KG-K W217018-20KG-K W217018-8KG-K
SIALW217018-1KG-KEA 74.8 EUR
SIALW217018-1KG-K SIALW217018-20KG-K SIALW217018-8KG-K
Methyl ethyl ketone, Sigma-Aldrich®
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6
Melting Pt: –86 °C
Density: 0.8054 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3 II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM