Acetone, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ300
W332607-1KG W332607-20KG W332607-8KG W332607-SAMPLE
SIALW332607-1KGEA 64.2 EUR
SIALW332607-1KG SIALW332607-20KG SIALW332607-8KG SIALW332607-SAMPLE
Acetone, Sigma-Aldrich®
Acetone
Formula: CH₃COCH₃
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM