Biuret, Sigma-Aldrich®
Supplier: Merck
15270-100G
15270-25G
SIAL15270-100GEA
233
EUR
SIAL15270-100G
SIAL15270-25G
Biuret, Sigma-Aldrich®
Biuret
Formula:
C₂H₅N₃O₂ Melting Pt: 188 °C Density: 1.467 g/cm³ (10 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007946 CAS Number: 108-19-0 EINECS: 203-559-0 Merck Index: 13,01305 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...