Ethyl acetoacetate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Acetoacetic acid ethyl ester, EAA
M-Clarity™ quality level = MQ200
688983-1KG 688983-240KG 688983-25KG
SIAL688983-1KGEA 120 EUR
SIAL688983-1KG SIAL688983-240KG SIAL688983-25KG
Ethyl acetoacetate, Sigma-Aldrich®
Ethyl acetoacetate
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 180.8
Melting Pt: –45 °C
Density: 1.0188 g/cm³ (20 °C)
Flash Pt: 65
Storage Temperature: Ambient
MDL Number: MFCD00009199
CAS Number: 141-97-9
EINECS: 205-516-1
Merck Index: 13,03793