Ethyl acetoacetate, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ300

W241504-1KG-K W241504-25KG-K W241504-5KG-K
SIALW241504-1KG-KEA 73.7 EUR
SIALW241504-1KG-K SIALW241504-25KG-K SIALW241504-5KG-K
Ethyl acetoacetate, Sigma-Aldrich®
Ethyl acetoacetate
Formula: C₆H₁₀O₃
Boiling Pt: 180.8 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.0188 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00009199
CAS Number: 141-97-9
EINECS: 205-516-1
Merck Index: 13,03793

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR