Ethyl acetoacetate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Acetoacetic acid ethyl ester, EAA
M-Clarity™ quality level = MQ300
W241512-100G-K W241512-1KG-K W241512-5KG-K
SIALW241512-100G-KEA 69 EUR
SIALW241512-100G-K SIALW241512-1KG-K SIALW241512-5KG-K
Ethyl acetoacetate, Sigma-Aldrich®
Ethyl acetoacetate
Formula: C₆H₁₀O₃
Boiling Pt: 180.8 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.0188 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00009199
CAS Number: 141-97-9
EINECS: 205-516-1
Merck Index: 13,03793