Sodium pyruvate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
P4562-100G P4562-25G P4562-5G
SIALP4562-100GEA 482 EUR
SIALP4562-100G SIALP4562-25G SIALP4562-5G
Sodium pyruvate, Sigma-Aldrich®
Sodium pyruvate
Formula: CH₃COCOONa
Melting Pt: 220…230 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4