Sodium pyruvate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
M-Clarity™ quality level = MQ200
P5280-25G
P5280-100G
P5280-500G
SIALP5280-25GEA
75.7
EUR
SIALP5280-25G
SIALP5280-100G
SIALP5280-500G
Sodium pyruvate, Sigma-Aldrich®
Sodium pyruvate
Formula:
CH₃COCOONa Melting Pt: 220…230 °C Density: 1.718 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002586 CAS Number: 113-24-6 EINECS: 204-024-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...