Diphenyl carbonate, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

D206539-100G D206539-1KG
SIALD206539-100GEA 34.1 EUR
SIALD206539-100G SIALD206539-1KG
Diphenyl carbonate, Sigma-Aldrich®
Diphenyl carbonate
Formula: C₁₃H₁₀O₃
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 78 °C
Density: 1.272 g/cm³ (14 °C)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00003037
CAS Number: 102-09-0
EINECS: 203-005-8
Merck Index: 12,07433

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR