Methoxyacetone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
177180-10G
177180-1G
SIAL177180-10GEA
209
EUR
SIAL177180-10G
SIAL177180-1G
Methoxyacetone, Sigma-Aldrich®
Methoxyacetone
Formula:
C₄H₈O₂ Boiling Pt: 118 °C (1013 hPa) Melting Pt: –66 °C Density: 0.957 g/cm³ (20 °C) Flash Pt: 24 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008768 CAS Number: 5878-19-3 EINECS: 227-549-0 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...