2,3-Pentanedione, Sigma-Aldrich®
Supplier: Merck
Synonyms: Pentane-2,3-dione
M-Clarity™ quality level = MQ100
241962-100G 241962-25G
SIAL241962-100GEA 109 EUR
SIAL241962-100G SIAL241962-25G
2,3-Pentanedione, Sigma-Aldrich®
2,3-Pentanedione
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 110
Melting Pt: –52 °C
Density: 0.96 g/cm³ (20 °C)
Flash Pt: 18
Storage Temperature: Refrigerator
MDL Number: MFCD00009313
CAS Number: 600-14-6
EINECS: 209-984-8
UN: 1224
ADR: 3 II