Acetonylacetone, Sigma-Aldrich®
Supplier: Merck
Danger
165131-100G 165131-25G
SIAL165131-100GEA 187 EUR
SIAL165131-100G SIAL165131-25G
Acetonylacetone, Sigma-Aldrich®
Acetonylacetone
Formula: C₆H₁₀O₂
Boiling Pt: 191 °C (1013 hPa)
Melting Pt: –5.4 °C
Density: 0.971 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00008792
CAS Number: 110-13-4
EINECS: 203-738-3
Merck Index: 13,00072