3,5-Heptanedione, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
280070-25G-A 280070-5G-A
SIAL280070-25G-AEA 199 EUR
SIAL280070-25G-A SIAL280070-5G-A
3,5-Heptanedione, Sigma-Aldrich®
3,5-Heptanedione
Formula: C₇H₁₂O₂
Boiling Pt: 47 °C (1013 hPa)
Density: 0.9464 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015186
CAS Number: 7424-54-6
EINECS: 231-054-5
UN: 1224
ADR: 3,III