Acetyl chloride 98%
Supplier: Thermo Fisher Scientific
Danger
151270010 151270025
ACRO151270010EA 70.3 EUR
ACRO151270010 ACRO151270025
Acetyl chloride 98%
Acetyl chloride
Formula: C₂H₃ClO
Boiling Pt: 50.9 °C (1013 hPa)
Melting Pt: –112 °C
Density: 1.1 g/cm³
Flash Pt: 4 °C
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 1717
ADR: 3,II
Merck Index: 13,00087
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3876 to 1.3896 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤30 APHA