Acetyl chloride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98%
CAS No.: 75-36-5
Molecular Formula: C2H3ClO
Formula Weight: 78.50
Flash Point: 4°(39°F)
Refractive Index: 1.3890
Odour: Pungent
Melting Point: -112°
Boiling Point: 50-52°
Beilstein Reference: 605303
Merck Reference: 14,85
UN No.: 1717
MDL No.: MFCD00000719
Formula:
C₂H₃ClO Boiling Pt: 50…52 °C Melting Pt: –112 °C Density: 1.104 Flash Pt: 4 °C (39 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000719 CAS Number: 75-36-5 EINECS: 200-865-6 UN: 1717 ADR: 3,II Merck Index: 14,85 |