Hexanoyl chloride, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

156957-100ML 156957-25ML
SIAL156957-100MLEA 65.3 EUR
SIAL156957-100ML SIAL156957-25ML
Hexanoyl chloride, Sigma-Aldrich®
Hexanoyl chloride
Formula: C₆H₁₁ClO
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: –87 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00000760
CAS Number: 142-61-0
EINECS: 205-549-1
UN: 2920
ADR: 8,II
Merck Index: 13,01769

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR