Hexanoyl chloride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
294659-100ML
294659-25ML
SIAL294659-100MLEA
239
EUR
SIAL294659-100ML
SIAL294659-25ML
Hexanoyl chloride, Sigma-Aldrich®
Hexanoyl chloride
Formula:
C₆H₁₁ClO Boiling Pt: 152 °C (1013 hPa) Melting Pt: –87 °C Density: 0.98 g/cm³ (20 °C) Flash Pt: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000760 CAS Number: 142-61-0 EINECS: 205-549-1 UN: 2920 ADR: 8,II Merck Index: 13,01769 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...