Valeryl chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Pentanoyl chloride, Valeroyl chloride
M-Clarity™ quality level = MQ200
94610-100ML 94610-500ML
SIAL94610-100MLEA 125 EUR
SIAL94610-100ML SIAL94610-500ML
Valeryl chloride, Sigma-Aldrich®
Valeryl chloride
Formula: C₅H₉ClO
Boiling Pt: 125 °C (1013 hPa)
Melting Pt: –110 °C
Density: 1.016 g/cm³ (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00000757
CAS Number: 638-29-9
EINECS: 211-330-1
UN: 2502
ADR: 8,II