Oxalyl dichloride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Oxalyl chloride
M-Clarity™ quality level = MQ200
221015-100G
221015-25G
221015-5G
SIAL221015-100GEA
121
EUR
SIAL221015-100G
SIAL221015-25G
SIAL221015-5G
Oxalyl dichloride, Sigma-Aldrich®
Oxalyl dichloride
Formula:
C₂Cl₂O₂ Boiling Pt: 63 °C (1013 hPa) Melting Pt: –12 °C Density: 1.48 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000704 CAS Number: 79-37-8 EINECS: 201-200-2 UN: 3129 ADR: 4.3,I Merck Index: 12,07046 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...