Oxalyl dichloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Oxalyl chloride
O8801-1KG O8801-100G O8801-2.5KG O8801-25G
SIALO8801-1KGEA 442 EUR
SIALO8801-1KG SIALO8801-100G SIALO8801-2.5KG SIALO8801-25G
Oxalyl dichloride, Sigma-Aldrich®
Oxalyl dichloride
Formula: C₂Cl₂O₂
Boiling Pt: 63 °C (1013 hPa)
Melting Pt: –12 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000704
CAS Number: 79-37-8
EINECS: 201-200-2
UN: 3129
ADR: 4.3,I
Merck Index: 12,07046