MULTIVIEW® Microarray Slides
Supplier: Enzo Life Sciences
MULTIVIEW®
ALEXADI9500200025EA 1090 EUR
ALEXADI9500200025 ENZOADI9500200025 ALEXADI9500200005 ENZOADI9500200005 ALEXADI9500200050 ENZOADI9500200050
MULTIVIEW® Microarray Slides
Slides Microarray Slides
These microarray slides with eleven human tissue specimens includes placenta, liver, tonsil, colon, skin, brain, breast (fibroadenoma), prostate, thyroid, kidney and fallopian tube.
  • High-consistency specimen and slide preparation for reduced maintenance
  • Fixed in formalin and paraffin embedded
  • Sections are placed on a SuperFrost Plus slide
  • Cores are approximately 2 mm in diameter
  • Tissue sections are 3 µm thick
Suitable for use with common FFPE tissue protocols as rapid screening tools for the optimisation or validation of stains in a broad-spectrum of immunohistochemical procedures.