Acetamide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
A0500-100G A0500-500G
SIALA0500-100GEA 53.2 EUR
SIALA0500-100G SIALA0500-500G
Acetamide, Sigma-Aldrich®
Acetamide
Formula: C₂H₅NO
Boiling Pt: 221.1 °C (1013 hPa)
Melting Pt: 78…81 °C
Density: 1.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008023
CAS Number: 60-35-5
EINECS: 200-473-5
Merck Index: 13,00044