Methylurea, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1-Methylurea, N-Methylurea, Methylcarbamide
M-Clarity™ quality level = MQ200
M86804-100G
M86804-500G
SIALM86804-100GEA
33.8
EUR
SIALM86804-100G
SIALM86804-500G
Methylurea, Sigma-Aldrich®
Methylurea
Formula:
C₂H₆N₂O Boiling Pt: 240 °C (1013 hPa) Melting Pt: 98…102 °C Density: 1.204 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007950 CAS Number: 598-50-5 EINECS: 209-935-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...