Acetanilide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
N-Phenylacetamide
M-Clarity™ quality level = MQ100
397237-1G
397237-5G
SIAL397237-1GEA
93
EUR
SIAL397237-1G
SIAL397237-5G
Acetanilide, Sigma-Aldrich®
Acetanilide
Formula:
C₈H₉NO MW: 135.17 g/mol Boiling Pt: 304 Melting Pt: 115 Density: 1.22 g/cm³ (20 °C) Flash Pt: 174 |
MDL Number:
MFCD00008674 CAS Number: 103-84-4 EINECS: 203-150-7 Merck Index: 13,00051 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...