Butyl butyrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Butyric acid butyl ester
M-Clarity™ quality level = MQ300
W218618-1KG-K W218618-9KG-K
SIALW218618-1KG-KEA 49.4 EUR
SIALW218618-1KG-K SIALW218618-9KG-K
Butyl butyrate, Sigma-Aldrich®
Butyl butyrate
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 166
Melting Pt: –91.5 °C
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009450
CAS Number: 109-21-7
EINECS: 203-656-8
UN: 3272
ADR: 3 III
Merck Index: 13,01555