Methyl laurate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester
M-Clarity™ quality level = MQ200
234591-2.5G 234591-25G 234591-5G
SIAL234591-2.5GEA 47.3 EUR
SIAL234591-2.5G SIAL234591-25G SIAL234591-5G
Methyl laurate, Sigma-Aldrich®
Methyl laurate
Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
Boiling Pt: 266
Melting Pt: 4.5
Density: 0.8694 g/cm³ (20 °C)
MDL Number: MFCD00008966
CAS Number: 111-82-0
EINECS: 203-911-3