Isoamyl acetate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Isopentyl acetate, Acetic acid isoamyl ester
M-Clarity™ quality level = MQ300
W205508-1KG-K W205508-20KG-K W205508-4KG-K
SIALW205508-1KG-KEA 73.7 EUR
SIALW205508-1KG-K SIALW205508-20KG-K SIALW205508-4KG-K
Isoamyl acetate, Sigma-Aldrich®
Isoamyl acetate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 142
Melting Pt: –78 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 25
Storage Temperature: Ambient
MDL Number: MFCD00008946
CAS Number: 123-92-2
EINECS: 204-662-3
UN: 1104
ADR: 3 III
Merck Index: 12,05125