Butyl acetate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ300
W217409-1KG-K W217409-20KG-K W217409-9KG-K
SIALW217409-1KG-KEA 77.5 EUR
SIALW217409-1KG-K SIALW217409-20KG-K SIALW217409-9KG-K
Butyl acetate, Sigma-Aldrich®
Butyl acetate
Formula: CH₃COO(CH₂)₃CH₃
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: –76 °C
Density: 0.875 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00009445
CAS Number: 123-86-4
EINECS: 204-658-1
UN: 1123
ADR: 3,III
Merck Index: 13,01534
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403329de943f8cb2ea6cd9d73ee35957d2140aa7&vid=682ab82de177b2985e94089408f7efebde59926d&prid=web&clid=SAPDAM