Dodecyl acetate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Lauryl acetate, Acetic acid Lauryl ester, Acetic acid Dodecyl ester

M-Clarity™ quality level = MQ300

W261602-1KG-K W261602-4KG-K
SIALW261602-1KG-KEA 151 EUR
SIALW261602-1KG-K SIALW261602-4KG-K
Dodecyl acetate, Sigma-Aldrich®
Dodecyl acetate
Formula: C₁₄H₂₈O₂
Boiling Pt: 265 °C (1013 hPa)
Density: 0.865 g/cm³ (25 °C)
MDL Number: MFCD00008973
CAS Number: 112-66-3
EINECS: 203-995-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR