Methyl formate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Formic acid methyl ester
M-Clarity™ quality level = MQ100
291056-100ML 291056-1L
SIAL291056-100MLEA 103 EUR
SIAL291056-100ML SIAL291056-1L
Methyl formate, Sigma-Aldrich®
Methyl formate
Formula: C₂H₄O₂
Boiling Pt: 32 °C (1013 hPa)
Melting Pt: –100.4 °C
Density: 0.9713 g/cm³ (20 °C)
Flash Pt: –32 °C
Storage Temperature: Ambient
MDL Number: MFCD00003291
CAS Number: 107-31-3
EINECS: 203-481-7
UN: 1243
ADR: 3,I
Merck Index: 12,06153