Methyl stearate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

W504807-10KG W504807-1KG W504807-5KG W504807-SAMPLE
SIALW504807-10KGEA 822 EUR
SIALW504807-10KG SIALW504807-1KG SIALW504807-5KG SIALW504807-SAMPLE
Methyl stearate, Sigma-Aldrich®
Methyl stearate
Formula: CH₃(CH₂)₁₆CO₂CH₃
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 1.007 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009005
CAS Number: 112-61-8
EINECS: 203-990-4
Merck Index: 12,08959

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR