Ethyl propionate
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Propionic acid ethyl ester
15498.A1
15498.D5
15498.A1EA
52.9
EUR
15498.A1
15498.D5
Ethyl propionate
Ethyl propionate
CAS No.: 105-37-3
Molecular Formula: CH3CH2CO2CH2CH3
Formula Weight: 102.13
Flash Point: 12°(54°F)
Physical Form: Liquid
Density: 0.891
Refractive Index: 1.3850
Melting Point: -73°
Boiling Point: 99°
Beilstein Reference: 506287
Merck Reference: 14,3847
UN No.: 1195
MDL No.: MFCD00009308
Formula:
C₅H₁₀O₂ Boiling Pt: 99 °C Melting Pt: –73 °C Density: 0.891 Flash Pt: 12 °C (54 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009308 CAS Number: 105-37-3 EINECS: 203-291-4 UN: 1195 ADR: 3,II Merck Index: 14,3847 |