Diethyl carbonate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
517135-100ML 517135-1L
SIAL517135-100MLEA 46.9 EUR
SIAL517135-100ML SIAL517135-1L
Diethyl carbonate, Sigma-Aldrich®
Diethyl carbonate
Formula: C₅H₁₀O₃
Boiling Pt: 127 °C (1013 hPa)
Melting Pt: –43 °C
Density: 0.9751 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009107
CAS Number: 105-58-8
EINECS: 203-311-1
UN: 2366
ADR: 3,III
Merck Index: 13,03815