Diethyl carbonate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
D91551-1L D91551-2.5L D91551-250ML D91551-18L D91551-4L
SIALD91551-1LEA 153 EUR
SIALD91551-1L SIALD91551-2.5L SIALD91551-250ML SIALD91551-18L SIALD91551-4L
Diethyl carbonate, Sigma-Aldrich®
Diethyl carbonate
Formula: C₅H₁₀O₃
Boiling Pt: 127 °C (1013 hPa)
Melting Pt: –43 °C
Density: 0.9751 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009107
CAS Number: 105-58-8
EINECS: 203-311-1
UN: 2366
ADR: 3,III
Merck Index: 13,03815