Dimethyl succinate ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A12565.0B
A12565.22
A12565.30
A12565.0BEA
120
EUR
A12565.0B
A12565.22
A12565.30
Dimethyl succinate ≥98%
Dimethyl succinate
Purity: 98+%
CAS No.: 106-65-0
Molecular Formula: C6H10O4
Formula Weight: 146.14
Flash Point: 85°(185°F)
Refractive Index: 1.4190
Melting Point: 18-19°
Boiling Point: 196°
Beilstein Reference: 956776
Merck Reference: 14,8869
MDL No.: MFCD00008466
Formula:
C₆H₁₀O₄ Boiling Pt: 196 °C Melting Pt: 18…19 °C Density: 1.119 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008466 CAS Number: 106-65-0 EINECS: 203-419-9 Merck Index: 14,8869 |