Diethyl oxalate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Oxalic acid diethyl ester

M-Clarity™ quality level = MQ200

135364-1KG 135364-25G 135364-3KG
SIAL135364-1KGEA 72.4 EUR
SIAL135364-1KG SIAL135364-25G SIAL135364-3KG
Diethyl oxalate, Sigma-Aldrich®
Diethyl oxalate
Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 185.7
Melting Pt: –41 °C
Density: 1.0785 g/cm³ (20 °C)
Flash Pt: 76
Storage Temperature: Ambient
MDL Number: MFCD00009119
CAS Number: 95-92-1
EINECS: 202-464-1
UN: 2525
ADR: 6.1 III
Merck Index: 13,03152

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR